SealPlate®, SealPlate® MiniStrips™, and ThinSeal™ Plate Sealing Film, Excel Scientific
Supplier: Excel Scientific
100-SEAL-PLT 100-THIN-PLT SP-2X8-50 SPS-2X8-50 STR-SEAL-PLT STR-THIN-PLT
490007-308BX 104.39 USD
490007-308 490007-312 490007-344 490007-360 490007-362 490007-366
SealPlate®, SealPlate® MiniStrips™, and ThinSeal™ Plate Sealing Film, Excel Scientific
Seals Microplate Seals
SealPlate® 50 µm thickness polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover.
SealPlate® MiniStripsTM sealing films are identical to SealPlate films above, but designed for sealing only one or two 8-well rows at a time on either strip-well or standard plates whenever rows must be selectively protected or accessed. Each MiniStrips film is 19.0 x 103.2 mm, including two positioning tabs. The long dimension with the tabs removed is 82.6 mm. MiniStrips are supplied in sheets of four from which strips canMore Product Information