Diversified Biotech Wash-N-Dry™ Slide Rack
Supplier: Diversified Biotech
WSDR-2000
490007-154EA 77.08 USD
490007-154
Diversified Biotech Wash-N-Dry™ Slide Rack
Slide Holders
Securely holds twelve slides for convenient cleaning, treating, staining and drying. Fits standard slide sizes including 25 mm & 1".
  • Offset design allows liquid and air to flow around and between all the slides ensuring best cleaning and faster drying times
  • Chemically resistant
  • Autoclavable