High Profile Orbital Shaker, Crystal Industries
Supplier: CRYSTAL TECHNOLOGY BE
SYC-2102A
490006-550EA 4850.92 USD
490006-550
High Profile Orbital Shaker, Crystal Industries
Shakers and Mixers Orbital Shakers
Orbital shakers can be used for various applications including culturing microbes, washing blots, and general mixing.  The High profile orbital shakers are designed and built for a heavy workload and provide continuous shaking with no vibration or heat output. Crystals orbital shakers can be placed into a standard incubator, converting the incubator into a shaking incubator.
  • Digital control and LED display
  • Low noise level
  • Gentle speed up and slow down
  • Automatic recovery from a power outage
Información accesorios: Platform accommodates: 20×25/50More Product Information