Dart® Daily Air Removal Test Pack, Steris
Supplier: Steris
NB113 NB215
48350-026CS 1520.22 USD
48350-026 48350-028
Dart® Daily Air Removal Test Pack, Steris
Sterilization Indicators
The Dart® daily air removal test is a pre-assembled Bowie-Dick test pack products used in monitoring healthcare steam sterilizers.
  • Lightweight, compact, one-piece system ready for use
  • Indicator strip can be kept as a permanent record
  • Lot number and expiration date on each test
  • Operates at 270-274°F/132-134°C with an exposure time of 3.5-4.0 minutes.
The daily air removal test is placed directly into an otherwise empty steam sterilizer chamber and does not require a retaining device. During processing, steam must displace the air through the blue heatMore Product Information