VWR® Premium Superfrost® Colorfrost® Microscope Slides
Supplier: VWR International
48311-950 18382-111 48311-601 48311-611 48311-656 48311-951 48312-401 48312-502 48312-604 48312-706 48382-101 48382-103 48382-119 75799-266
48311-950CS 685.9 USD
48311-950 18382-111 48311-601 48311-611 48311-656 48311-951 48312-401 48312-502 48312-604 48312-706 48382-101 48382-103 48382-119 75799-266
VWR® Premium Superfrost® Colorfrost® Microscope Slides
Slides Microscope Slides
VWR® Superfrost® and Colorfrost® Micro Slides feature brilliant coloring and a coating that accepts and retains laboratory markings better than comparable products.
  • Easy to write on, easy to read
  • Key shows which side is up
  • Eliminate sticking and scratching
Slides are frosted and have a brilliant colored, opaque coating
All slides have the word “SPECIMEN” printed on them, except for the white Premium Superfrost slide, which is unprinted.
All slides are 75x25mm and are autoclavable.
Coating takes and retains most laboratory marking methodsMore Product Information