LiChrolut® Polypropylene Extraction Columns, MilliporeSigma
Supplier: MilliporeSigma
1.19847.0001 1.19849.0001 1.19855.0001 1.02014.0001 1.02014.0001 1.19127.0001 1021220001
48219-204PK 534.92 USD
48219-204 48219-216 48219-220 48219-231 48219-232 48219-234 48219-236 48219-242 48219-246 EM1.02014.0001 10143-296 10143-778 10144-030 10144-032 10144-036
LiChrolut® Polypropylene Extraction Columns, MilliporeSigma
Chromatography Columns
For nonpolar extraction, choose RP-18, RP-18E (Endcapped), or CN (Cyano) columns.
Typical sample matrix: aqueous buffer solution. Typical sample substances: aromatic ring systems and compounds with alkyl chains. Typical elution solvent: acetonitrile, methanol, and ethyl acetate. Endcapped cartridges have a bonded phase that is reacted with trimethylchlorosilane to cover silanols not previously reacted with the initial bonding reagent. For polar extraction, choose CN (Cyano) or Si (Silica Gel) columns. Typical sample matrix: hexane, oils, and chlorinated hydrocarbons. Typical sampleMore Product Information