SP Bel-Art CryoSafe Mini Quick-Freeze Cooler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18846-0010
47751-728EA 239.86 USD
47751-728
SP Bel-Art CryoSafe Mini Quick-Freeze Cooler, Bel-Art Products, a part of SP
Coolers Benchtop Coolers
This CryoSafe mini quick-freeze cooler can be used for up to 45 minutes to rapidly and repeatedly chill biological samples for use in molecular biology.
  • Holds twelve 0.5 or 1.5 ml microcentrifuge tubes
  • No direct contact between the tubes and the insulating solution; labels and markings are safe
  • Clear polycarbonate lid shows positions of tubes and is keyed to correspond with positions printed on cooler
  • Molded side handles allow secure grip for transfer
  • Rubber feet to prevent sliding on the lab bench
  • Stackable
Applications such as ethanol precipitationMore Product Information