ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: Eppendorf
0030078535 0030078543 0030078578 0030078594 0030078500 0030078519 0030078527 022491288 0030078560 0030078616 0030078551 0030078624 0030078810 0030078829 0030078845 0030078586
47745-090CS 205.41 USD
47745-090 47745-092 47745-096 89126-924 47745-084 47745-086 47745-088 47747-994 47745-094 89132-036 89014-309 75845-010 76018-992 76018-994 76018-998 76423-882
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Pipette Tips
Eppendorf ep Dualfilter T.I.P.S.® Racks offer 'PCR clean and Sterile' grade filter pipette tips, providing dual-layer protection against contamination for critical lab applications, ensuring precision, reliability, and hygiene in high-stakes environments.
  • Two filter layers provide dual protection against aerosols and biomolecules to guard both pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent
More Product Information