SP Bel-Art Magnetic Stirring Bars, PTFE, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
37119-9003
47744-998CS 492.1 USD
47744-998
SP Bel-Art Magnetic Stirring Bars, PTFE, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Stir bar with removable stabilizing ring to keep it centered in vessel.
  • Removable stabilizing ring
  • PTFE construction
  • Durable
  • Resists spinout
Creates deep vortex without splashing or chattering and resists spinout.