Eisco® 4-Piece Slotted Mass Set with Hanger, Stainless Steel
Supplier: Wards
PH0258MSS
470344-716EA 31.6 USD
470344-716
Eisco® 4-Piece Slotted Mass Set with Hanger, Stainless Steel
Weights Calibration Weights
Set of 4 Slotted Masses with a hanger, made of stainless steel and engraved for identification.
  • Includes 4 individually calibrated 50 g slotted masses and a 50 g mass hanger with hook
  • Total weight of set is 250 g
  • Made of high-quality polished stainless steel
  • Each piece is engraved to identify its mass
  • Good classroom tool for demonstrations on force and balance
Slotted Mass Set comprised of four individual 50g masses and one 50 g hanger. Made of high-quality, highly polished stainless steel. Each piece is engraved to identify its mass value. Masses areMore Product Information