Density Cubes with Hook

Supplier: Wards
PH0108K PH0108N PH0108J
470343-818EA 6.29 USD
470343-818 470344-624 470345-220
Density Cubes with Hook
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Cubes have 2 cm(0.78") sides with a hook for density investigation and specific gravity activities. Element stamp indicates material.

  • Single, solid density cube with hook and stamped with element for easy identification
  • Measures 2 cm(0.78") cubed, including the hook, the cube measures 1.5" in height
  • Excellent for physics classrooms
  • Density Cubes have a wide range of educational applications for all grade levels, including density investigation, specific heat and specific gravity exploration

Since these cubes are equipped with hooks, they can be easilyMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR