Wash Bottle for Water
Supplier: Wards
CHWB1020A
470344-058EA 5.19 USD
470344-058
Wash Bottle for Water
Bottles Wash Bottles
Wash bottle for water with color-coded labels, wide mouth, self-venting design. Bottle is made of durable polypropylene, ideal for laboratory use with easy and precise dispensing.
  • Easy to read labeling, including chemical information and precautionary statements
  • Blue cap with built-in nozzle
  • Wide mouth for easy filling
  • Self venting design to eliminate leaks and dripping
  • Easy to control squeeze for dispensing
Made of low density polyethylene (LDPE) for superior quality and strength. Innovative flex ink won't fade, crack, or peel. Bottle is labeled forMore Product Information