Ward's® Pipette Tip Disposal Box
Supplier: VWR International
470311-178
470311-178EA 93.79 USD
470311-178
Ward's® Pipette Tip Disposal Box
Boxes Waste Disposal Boxes
Quick flip of the port cover makes tip ejection easy.
  • Large, hinged lid allows easy disposal of contents
  • 3/8" (10 mm) thick acrylic provides protection from Beta radiation
The ward's pipette tip disposal box is convenient for one-handed disposal of used pipette tips, and eliminates any need to handle pipette tips used with hazardous fluids. Biohazard disposal bags can be used in conjunction with this box if hazardous fluids are not acrylic friendly.