Suture Training Sheets
Supplier: WALLCUR, LLC.
REM-TIS-SKIN-01L
470306-756EA 245.39 USD
470306-756 470306-794
Suture Training Sheets
Models Human Biology Models
Multiple layers of closure practice.
  • Use alone, or on other suture training frames
  • Available in basic or advanced styles
  • Each layer reacts differently to closure
The Basic Tissue sheet is for use in the practice of basic closure methods. The Tissue plate is made of 4 layers developed to match the feel of epidermis, dermis, fat, and muscle. The Advanced Tissue Insert is for use in the practice of all common closure methods. The tissue plate is made of 5 layers developed to match the feel of epidermis, dermis, fat, muscle andMore Product Information