Ammonium Acetate

Supplier: ALDON CORP SE

AA0219-500ML
470300-176EA 17.05 USD
470300-176
Ammonium Acetate
Chemicals for Science Education

CAS Number: 631-61-8
Formula Weight: 77.09
Formula: CH3COONH4
Hazard Info: Irritant
Density (g/mL): 1.07
Boiling Point (°C): Decomposes
Freezing Point (°C): 114
Solubility: Water and Alcohol
Synonyms: Acetic Acid Ammonium Salt
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Información de suministro: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR