Ward's® Sterile Cell Strainers
Supplier: VWR International
470236-274 470236-276 470236-278 470236-280 470236-282 470236-310 470236-312 470236-314
470236-274EA 62 USD
470236-274 470236-276 470236-278 470236-280 470236-282 470236-310 470236-312 470236-314
Ward's® Sterile Cell Strainers
Cell Strainers
Ward's® Sterile Cell Strainers and Cell-Straining Kits provide all the necessary parts for filtration of cell suspensions.
  • Available as Kits (strainer, 50 ml tube, and cap) or as strainers alone
  • Easy to open under the hood, reducing contamination risk
Cell Straining Kits: Eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean users are no longer forced to expose entire bags of 50 ml tubes and corresponding caps in order to performMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable