GSC International Conductivity of Solutions Demonstration
Supplier: GSC International
4-31001 4-31001-10
470002-034EA 18.45 USD
470002-034 470199-910 470235-678
GSC International Conductivity of Solutions Demonstration
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Innovative Device Makes Visualizing Conductivity Possible.
  • Much safer than 110 V conductivity meters
  • Students can clearly see which solutions conduct electricity
  • Reusable year after year
  • Instructions available for download
Observe increases and decreases in a liquids conductivity with this innovative apparatus and demonstration. A molded socket on a PVC cover is provided with binding posts for connecting two wire cords. Two electrodes are also internally connected to the lamp circuit. Fit the molded cover over the kits glass tumbler filled with aMore Product Information