Tube-CUBE™ Specimen Tube Blocks
Supplier: United Product & Instrument
54900 54400
470229-864EA 68.35 USD
470229-864 470229-866
Tube-CUBE™ Specimen Tube Blocks
Racks Tube Racks
The Basis For Sample Management In Any Type Of Laboratory.
  • Invert and pour! - tubes will stay in place
  • Easily disinfected if contaminated, great for phlebotomy trays
  • Organize and store samples in refrigerator or freezer
The do all mini racks! Each of these Tube storage racks conveniently fits into the same footprint, making it easy to plan your storage of samples in confined spaces. Each of the trays is designed to fit neatly into a phlebotomy tray, or easily into carts. The tube slots are all flanged to hold the tubes securely, preventing spills andMore Product Information