TeacherGeek Judo Bot
Supplier: TEACHERGEEK INC
1824-72 1824-62
470229-102EA 13.48 USD
470229-102 470229-104
TeacherGeek Judo Bot
Educational Materials Physics Educational Materials TeacherGeek System Components
Get ready to R-R-RUMBLE!!
  • Utilize the Design Engineering Process
  • Encourage creativity, critical thinking and problem solving
  • Learn about push and pull forces, wind, and sail design
  • Apply the scientific method
Use the power of hydraulic cylinders to move your Judo-Bot up, down, and side-to-side to tip or push your opponent out of the ring. An overview document guides you through this STEM activity. Start by learning about hydraulic and pneumatic power in the Fluid Power Lab. Then, design your own Judo-Bot to compete in Design and Engineering Challenges.More Product Information