NeuLog WiFi Module
Supplier: Wards
WIFI202
470221-194EA 219.2 USD
470221-194
NeuLog WiFi Module
Data Loggers
Wi-Fi simplifies data acquisition.
  • Compatible with iPads, tablets, and PCs
  • No downloads or installations
  • Compatible with popular browsers
The Wi-Fi module is one of the most advanced and innovative elements of NeuLog. Using Wi-Fi technology, students can take measurements with NeuLog on nearly any kind of device including a tablet with Android, a PC computer with Windows or Linux, or a Macintosh with OS.
These devices communicate with the Wi-Fi adaptor for a unique experience of data acquisition. Users don’t need to download any applications orMore Product Information