Universal Mirror
Supplier: Wards
PH0528
470219-444EA 10.45 USD
470219-444
Universal Mirror
Mirrors
With this stainless steel mirror, students can perform experiments using light rays with plane, concave, and convex mirrors all in one convenient and easy to use tool. The flexible metal curve can be switched to convert from a concave to convex mirror.