Compound Bar
Supplier: Wards
PH0384B
470219-434EA 18.3 USD
470219-434
Compound Bar
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
For demonstrating the differential expansion of two metals as shown by the curvature produced when the bar is heated. Each metal band is 200 x 15mm and riveted together.
Order Now
SPECIFICATIONS
Environmentally Preferable