Compact Electrophoresis Power Supply
Supplier: Thermo Scientific
105ECA-115
470204-250EA 1290.65 USD
470204-250
Compact Electrophoresis Power Supply
Power Supplies
This compact power supply is ideal for DNA or RNA electrophoresis.
  • Easy-to-use power supply operates in constant voltage
  • Adjustment knob and hi-lo range switch control the output, which is variable in 10 V increments from 20 V to 250 V
  • Two-position switch toggles the bright LED display from indicating output voltage to output current
  • Equipped with an overcurrent detector for maximum safety
Carry out DNA and RNA electrophoresis with the help of the Owl EC-105 Compact Power Supply, equipped with an overcurrent detector for maximum safety. Unit has twoMore Product Information