Basic Protist Survey Slide Set
Supplier: Avantor
470183-454EA 42 USD
470183-454
Basic Protist Survey Slide Set
Slides Prepared Slides
Representatives from four Phyla
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
All slides are prepared as whole mounts.
Información para pedidos: Set includes Chlorophyta: Spirogyra, vegetative; Chrysophyta (Stramenopila): Diatoms, mixed; Ciliata (Alveolata): Paramecium; Euglenozoa: Euglena; Rhodophyta: Bactrachospermum; and Sarcodina (Rhizopod): Amoeba.