Ward's® Onion (Allium) Slides
Supplier: Avantor
470176-874EA 14.85 USD
470176-874 470176-876 470177-830 470177-452 470177-454 470177-456 470181-228 470182-532 470182-536
Ward's® Onion (Allium) Slides
Slides Prepared Slides
Great example of plant cells and mitosis.
  • Ward's produced slides
  • Rigorous quality control standards
Teach students about mitosis in plant cells with any of the several onion mitosis slides available. Each slide shows all stages of mitosis. One size doesn't fit all, so there are many different views of the tissue, stained with a variety of compounds to enhance various structures. Most teachers prefer a longitudinal section through the root tip. Cross sections provide an additional perspective of mitosis in the cell. Epidermal peels are the best
More Product Information