Ward's® Sedimentary Rock Collection
Supplier: Avantor
470175-706EA 175.49 USD
470175-706 470025-224
Ward's® Sedimentary Rock Collection
Specimens Rock Specimens Sedimentary Rocks
Represents a Wide Range of Depositional Environments and Rock Textures
  • Basic introduction to organic material
  • Compartmented box
Use this set as a basic introduction to rocks formed from organic material or fragments of other rocks. The set is housed in a compartmented collection box and comes with an identification list.
Contents:
12 numbered samples, 1" x 2"
Rocks:
chert, conglomerate, coquina, gypsum, limestone (2), sandstone (2), shale (2), siltstone, travertine
Hand Specimen Set
Large hand samples are ideal for demonstration and display. This set isMore Product Information