Sterilizing Tote Boxes
Supplier: Dynalon
410545
470173-916CS 310.65 USD
470173-916
Sterilizing Tote Boxes
Boxes ESD-Safe Boxes
This lighter-weight polypropylene tote is easier to carry and can even be autoclaved. Chemically resistant to acids, alkalis, oils, and detergents, it will not rust or corrode. They can be nested for compact storage.