Utility Jar
Supplier: Dynalon
600335
470173-740CS 652.85 USD
470173-740
Utility Jar
Jars General Use Jars
One gallon capacity, high density polyethylene utility jar with metal closure.
  • Wide mouth, natural color
  • Withstands temperatures to 110 °C
Hundreds of uses around the lab.