Kimwipes
Supplier: Kimberly-Clark
34256
470173-498BX 19 USD
470173-498
Kimwipes
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
Dependable, highly absorbent tissues.
  • Specially designed for lab use
  • Absorb up to 15 times their weight in liquid
Perfect for polishing glassware or cleaning instruments because they will not scratch or deposit lint. Supplied in handy, pop-up dispensing cartons.
Información para pedidos: Now available in handy convenience packs. Great for classrooms!
Order Now
SPECIFICATIONS
Environmentally Preferable