Heat Conductometer
Supplier: Shiv Dial Sud & Sons
P20279A
470148-760CS 392.5 USD
470148-760
Heat Conductometer
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This unit visually reinforces heat conductivity lessons.
  • Five metals included: aluminum, brass, copper, nickel, andsteel
  • Instructions included
It demonstrates how five different metals—aluminum, brass, copper, nickel, and steel—conduct heat. The five rods are heated over a flame simultaneously, and students gauge what metal conducts heat best by observing how fast the wax at the tip of each rod melts.
Información para pedidos: The apparatus comes with instructions. Wax and a heat source are required but not included.