Ward's® Ballistics Flashcards
Supplier: PROFORMA ASSOCIATES
323382
470147-538BX 8394.8 USD
470147-538
Ward's® Ballistics Flashcards
Educational Materials Forensics Educational Materials Crime Scene Evidence Learning Activities
An Excellent Reference of Real Discharged Bullets and Casings
  • Beautiful Images
  • Students Can Drill Themselves for Exams
Observe lands and grooves around the entire circumference of bullets with our exclusive flashcards. Images of four separate bullets are provided in order to make comparisons, in addition to images of the breechface of four shell casings, which show the firing pin impression on the primer of the casing. Size: 3" x 5". Set of 20.
Order Now
SPECIFICATIONS
Environmentally Preferable