Fundamentals of Physics Newton Cart
Supplier: Wards
NCWR-11
470128-706EA 146.2 USD
470128-706
Fundamentals of Physics Newton Cart
Educational Materials Physics Educational Materials Mechanics Learning Activities
Hands-On and Versatile Demonstration of Mechanics
  • Includes a text with activities designed for high school level study
  • Unit is approximately 39" in height
This gravity-powered metal car demonstrates the various principles of mechanics, including Newton's Laws of motion, energy and energy conversion, momentum, impulse, and work, showing each concept individually as well as integrated together in a system. As energy weights fall from the cart, the force of the weight produces tension in a flexible driveline. The car accelerates at a constant rate untilMore Product Information