Lab-Aids® Human Genetics Experiment Kit
Supplier: Lab-Aids
7
470111-670EA 65.1 USD
470111-670
Lab-Aids® Human Genetics Experiment Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
How are inherited traits transmitted?
  • Develop an experiment
  • Understand and demonstrate principles of heredity
  • Time required: one 50 minutes class period
The experiment is based on the transmission of a single inherited trait - the ability to taste harmless PTC (phenylthiocarbamide). The goal is for students to determine if they have this gene. They collect data by testing family members, correlating and pooling the data for the entire class, then calculating the random segregation data according to the Hardy-Weinberg Law. Background information aboutMore Product Information