Bocce Ball Pendulum
Supplier: INDUSTRIAL SUPPORT INC.
180209
470039-722EA 75 USD
470039-722
Bocce Ball Pendulum
Educational Materials Physics Educational Materials Mechanics Learning Activities
Large scale demonstration of harmonic motion.
  • Large scale demonstration
  • Easily attaches to any ceiling
Help students see the answers to conservation of energy, vectors, and tension questions are right in front of their faces. This classical experiment has been adapted to excite, engage, and intrigue students. A 5 lb. bocce ball is hung from a 25 ft. long rope with a breaking point of 500 lbs. Students hold this ball to their nose and release it, with the goal to stay still as it returns just short of its initial position. For extra stability, the ropeMore Product Information