Shower Flush System
Supplier: JOEL C. PALMER
175710
470039-360EA 154.8 USD
470039-360
Shower Flush System
First Aid Supplies Showers
Perform the monthly shower flushing necessary to Comply with OSHA standards with this device.
  • An innovative water catcher for flushing the emergency chemical shower
Because many lab drains are not ideally located or equipped to contain all the water released during an emergency flushing, this system ensures the water flows at a steady pace that all drains can easily accommodate.
An excellent complement to the shower tester (Wards® 64990-03), this shower flush system includes an elbow valve, flex hose, drain cover and hole cutter for the garbage canMore Product Information