Ward's® Quartz (Chert)
Supplier: Avantor
470025-756EA 19.69 USD
470025-756 470025-758 470025-760
Ward's® Quartz (Chert)
Specimens Mineral Specimens Classroom Minerals
Light gray, compact, opaque, microcrystalline.
Información para pedidos: WARD’S offers different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces are suitable for classroom study, while the larger specimens in the one kilogram pack can be cut toMore Product Information