Ward's® Introductory Mineral Collection
Supplier: Avantor
470025-258EA 42.29 USD
470025-258
Ward's® Introductory Mineral Collection
Specimens Mineral Specimens Mineral Collections
Beginner’s Collection Illustrates Important Physical Properties
  • Cut by WARD's Geologists
  • Highest Quality Specimens
  • Great for Introducing Students to Minerals
Color, hardness, luster, cleavage, streak, common habit, and specific gravity are all represented in one compact, economical set of 12 minerals. Beginning geology students will easily recognize each of the wide range of physical properties when they examine the basic minerals. The set is housed in a compartmented collection box, and an identification list is included.
Contents:12 numberedMore Product Information