Ward's® Electrophoresis Techniques Reference Poster
Supplier: Avantor
470023-282EA 31.99 USD
470023-282
Ward's® Electrophoresis Techniques Reference Poster
Charts
Clearly Presents Guidelines for Agarose and PAGE Procedures
  • All key steps and applications of gel electrophoresis
  • Excellent wall reference
  • Artfully illustrated
Students will always know which step comes next when our exclusive poster is on display in the lab. Agarose electrophoresis for DNA analysis and polyacrylamide gel electrophoresis (PAGE) for protein analysis are both detailed in a step-by-step format, highlighted with photographs and illustrations that show how to cast, load, run, stain, and distain a gel. It also features information on theMore Product Information