Student Plankton Net
Supplier: Peco Sales
77919
470021-296EA 299.99 USD
470021-296
Student Plankton Net
Samplers Water Samplers
Perfect for Introductory Plankton Sampling
  • Economical
This 20 1/2" Nitex plankton net is attached to a 6" diameter stainless steel ring and bridle. A clear, polycarbonate cod-end assembly with latex tube and pinch clamp and a ten meter calibrated polypropylene line are also included.