Ward's® Tuff (Rhyolitic)
Supplier: Avantor
470015-516EA 21.39 USD
470015-516 470026-308 470219-930
Ward's® Tuff (Rhyolitic)
Specimens Rock Specimens Igneous Rocks
Gray, fine, friable, with abundant small sanidine crystal fragments.
  • Student, hand, or bulk specimens available
Ward's® offers different specimen sizes to meet your various needs:
Student Specimens: Ideal for students to use for study and identification. Specimens range in size from 1" x 1" to 1" x 2" and come in packages of ten.
Hand Specimens: Great for lectures, demonstrations, and display. Specimens range in size from 2" x 3" to 3" x 4", and each comes in a labeled plastic tray.
Bulk Pack: Several of the smaller pieces are suitable forMore Product Information