Ward's® Agarose Gel Electrophoresis Lab Pack
Supplier: Avantor
470015-288EA 64 USD
470015-288
Ward's® Agarose Gel Electrophoresis Lab Pack
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Bundled for Convenience
  • Materials for 6 lab groups
The kit includes one 500 mL bottle of 5 X TBE buffer concentrate, 5 g of ultra-pure agarose, one 60 mL bottle of Ward's QUIKView DNA Stain, 5 mL of 10X loading dye and a staining tray.