Precision Photoelectric Effect
Supplier: The Science Source
EP-07
470006-464EA 1824.95 USD
470006-464
Precision Photoelectric Effect
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Precision Photoelectric Effect uses three color filters to calculate Planck's Constant.
  • Three Filters of Wavelength Included
  • Students Use the Graph's Slope to Calculate Planck's Constant
  • Apparatus Shows that Photoelectrons' Energy Relies on Wavelengths of Incoming Radiation
The Photoelectric Effect demonstrates that the energy of a photoelectron depends on the wavelength of the incoming radiation and not on its intensity. As monochromatic radiation irradiates the photocathode, a potential is applied opposing the energy of the emitted electrons.More Product Information