Clear Polystyrene Spot Plates
Supplier: PRECISION MOULDED POLYMERS LTD
AM9000.00000.00000
470005-972EA 21 USD
470005-972
Clear Polystyrene Spot Plates
Staining Tiles
Offering more depressions than other spot plates, these plates are suitable for most lab applications. Made of clear polystyrene with an attached measuring scoop.
  • 15 depressions measure 2 ml deep
  • Each plate measures 140×80 mm with 15 depressions
Envase: Package of 10.
Precaución: Not recommended for use with strong solvents.