Solar Cell
Supplier: Wards
PH1320SPL
470002-992EA 62.7 USD
470002-992
Solar Cell
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Show How Direct/Indirect Sunlight Provides Power
A photo-voltaic cell, this solar cell uses illumination to produce a voltage able to drive current through a load. The cell used in this device is incredibly sensitive and capable of delivering 100mW into a 10 load when a 36W filament bulb is held approximately 30 mm from the cell.
Order Now
SPECIFICATIONS
Environmentally Preferable