Protein InstaStain®
Supplier: Edvotek
2016
470002-532EA 84.7 USD
470002-532
Protein InstaStain®
Chemicals for Science Education
High Quality Results, Excellent for Photodocumentation
Stain protein gels in just 20 minutes, plus fixing and destaining times. Protein InstaStain® is far faster than liquid blue stains and ensures a yield of uniform gel results. Environmentally friendly InstaStain® also uses a solid matrix to avoid large amounts of liquid waste. For 15 gels.