Anticon® Presaturated Wipes
Supplier: Contec
495357-660
45800-088CS 504.55 USD
45800-088 45800-084
Anticon® Presaturated Wipes
Wipes Disposable Wipes and Towels Presaturated Wipes
Anticon® Presaturated Wipes are manufactured using ultra-clean Anticon® Gold and other Anticon® wipes in conjunction with semiconductor or USP grade solvents.
  • Wipes 45800-088 and 45800-084 have a volume of 375 ml per bag
  • Wipe 89092-960 has a volume of 60 ml per bag
These polyester knit cleanroom wipes are presaturated with the optimum amount of solution to deliver absolute cleaning power, not excess solution.
Anticon® presats are delivered with a patented low particle generation technology. Use Anticon® presats with the knowledge and security thatMore Product Information