Tantalum (Ta) Lumina™ Hollow Cathode Lamp, PerkinElmer
Supplier: PerkinElmer
N3050177
45603-354EA 1610.31 USD
45603-354
Tantalum (Ta) Lumina™ Hollow Cathode Lamp, PerkinElmer
Lights Hollow Cathode Lamps
Single element Lumina™ hollow cathode lamp (HCL) for the detection of elemental Tantalum (Ta). Series N30501XX Lumina™ 2" (50 mm) diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that this element requires the use of a nitrous oxide-acetylene flame. Every genuine , PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determiningMore Product Information