Corning® Tris, 10X Buffered Saline, Liquid, Corning
Supplier: Corning
46-012-CM
45001-132PK 397.51 USD
45001-132
Corning® Tris, 10X Buffered Saline, Liquid, Corning
Buffers Cell Culture Buffers
Tris Buffered Saline (TBS) is an isotonic, non-toxic buffer used in some biomedical techniques to maintain pH within a relatively narrow range
TBS can shift pH relatively dramatically as temperature changes, with the pKa declining approximately 0.03 units per degree Celsius rise in temperature.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage Temperature Ambient
Shipping Temperature Ambient