Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
21-031-LX 55-031-PC 21-031-LB 20-030-CV 20-031-CV 21-030-CV 21-030-CM 21-031-CV 21-031-CM
45000-438EA 354.89 USD
45000-438 45000-444 45000-440 45000-442 45000-426 45000-428 45000-430 45000-432 45000-434 45000-436
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Buffers Cell Culture Buffers
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Complete Certificate of Analysis available for each production lot, along with full formulation information.
It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Precaución: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C