Slimline™ Flathead Mop Covers and Hardware, Micronova
Supplier: Micronova
SLSC6-18IR SLSC6-14IR SLSC6-9IR SLSC6-9 SLSC6-14 SLSC6-18 SLSC6AA-14 SMC6-24B SMC6-30B
43300-966CS 335.78 USD
43300-966 43300-968 43300-970 11310-602 11310-604 11310-606 94023-842 75845-734 76323-720
Slimline™ Flathead Mop Covers and Hardware, Micronova
Mops
A series of elasticized slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitizing all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
  • Covers are easy to change and fit securely over mop frame
  • Fully encased Urethane Foam layer for ample disinfectant dispersal
  • Single-ply covers available without foam interior
  • Fits on QPSL plastic frame or QDSL stainless steel frame
  • Suitable for use with strong disinfectants and repeated autoclaving
The MicronovaMore Product Information